Assay Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009701.1 | NP_001009701.1 | 844 | 145 | ||
| GenBank mRNA | BC088404.1 | - | 844 | 7108395 | 145 | |
More Information
Set Membership: |
|

Itfg3
Genomic Map