Assay Details
| Gene Symbol: Arf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080904.2 | NP_543180.1 | 2 - 3 | 334 | 154 | |
| XM_006257320.2 | XP_006257382.1 | 3 - 4 | 543 | 154 | ||
| GenBank mRNA | AY325223.1 | - | 3 - 4 | 301 | 154 | |
| BC088865.1 | - | 2 - 3 | 334 | 7319260 | 154 | |
| L12382.1 | - | 3 - 4 | 447 | 154 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Arf3
Genomic Map