Assay Details
| Gene Symbol: Oas1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009379.1 | NP_001009379.1 | 1064 | 68 | ||
| GenBank mRNA | AY196699.1 | - | 1064 | 68 | ||
| Gene Symbol: LOC103689988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769299.1 | XP_008767521.1 | 5 - 6 | 1164 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Oas1d
Genomic Map