Assay Details
| Gene Symbol: Oas1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009490.1 | NP_001009490.1 | 4 - 5 | 923 | 136 | |
| GenBank mRNA | AY196700.1 | - | 4 - 5 | 923 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oas1f
Genomic Map