Assay Details
| Gene Symbol: Oas1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009492.1 | NP_001009492.1 | 3 - 4 | 697 | 165 | |
| XM_008769248.1 | XP_008767470.1 | 3 - 4 | 698 | 165 | ||
| GenBank mRNA | AY196698.1 | - | 3 - 4 | 697 | 165 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oas1e
Genomic Map