Assay Details
| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009718.1 | NP_001009718.1 | 2 - 3 | 236 | 137 | |
| GenBank mRNA | AY651018.1 | - | 2 - 3 | 236 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klra2
Genomic Map