Assay Details
| Gene Symbol: Klra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009718.1 | NP_001009718.1 | 5 - 6 | 683 | 160 | |
| XM_008763403.1 | XP_008761625.1 | 4 - 5 | 745 | 160 | ||
| GenBank mRNA | AY651018.1 | - | 5 - 6 | 683 | 160 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klra2
Genomic Map