Assay Details
| Gene Symbol: Kbtbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012045.2 | NP_001012045.2 | 1 | 1608 | 85 | |
| GenBank mRNA | BC086326.1 | - | 1447 | 7114652 | 85 | |
| Gene Symbol: Kbtbd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252370.2 | XP_006252432.1 | 1870 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kbtbd6
Genomic Map