Assay Details
| Gene Symbol: Ptgdr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012070.1 | NP_001012070.1 | 1 | 1150 | 150 | |
| XM_006231040.2 | XP_006231102.1 | 1 | 1238 | 150 | ||
| XM_008760231.1 | XP_008758453.1 | 2 | 1216 | 150 | ||
| GenBank mRNA | AY228550.1 | - | 1 | 1150 | 150 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ptgdr2
Genomic Map