Assay Details
| Gene Symbol: Mtmr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012077.1 | NP_001012077.1 | 14 - 15 | 1590 | 157 | |
| XM_006232046.2 | XP_006232108.1 | 14 - 15 | 1681 | 157 | ||
| GenBank mRNA | BC089876.1 | - | 14 - 15 | 1590 | 7325487 | 157 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P061725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115374 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P061725 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P061725 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P061725 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100523 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17555 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7644 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62004 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10813759 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813769 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813768 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813766 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813756 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813764 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813765 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813762 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813761 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813758 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813757 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813763 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813767 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813755 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813760 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10813752 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200003736.rn.1 | TC0200000588.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200003735.rn.1 | TC0200000588.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10813752 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21862398 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376273 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375691 | - | - |

Mtmr12
Genomic Map