Assay Details
| Gene Symbol: Ang2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012359.1 | NP_001012359.1 | 1 | 440 | 116 | |
| XM_006251898.2 | XP_006251960.1 | 3 | 832 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ang2
Genomic Map