Assay Details
| Gene Symbol: Rnase10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012467.1 | NP_001012485.1 | 1 | 564 | 117 | |
| XM_006251868.2 | XP_006251930.1 | 2 | 2071 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnase10
Genomic Map