Assay Details
Gene Symbol: Vom2r31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173319.1 | NP_775441.1 | 3 - 4 | 1311 | 132 | |
GenBank mRNA | AF016184.1 | - | 3 - 4 | 1311 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
