Assay Details
| Gene Symbol: tGap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007635.2 | NP_001007636.2 | 16 - 17 | 2290 | 135 | |
| XM_006232134.2 | XP_006232196.1 | 18 - 19 | 2632 | 135 | ||
| GenBank mRNA | AY631396.1 | - | 16 - 17 | 2290 | 135 | |
| AY631398.1 | - | 9 - 10 | 1434 | 135 | ||
| AY631399.1 | - | 16 - 17 | 2055 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

tGap1
Genomic Map