Assay Details
| Gene Symbol: Krt28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008760.1 | NP_001008760.1 | 2 - 3 | 518 | 109 | |
| XM_006247356.2 | XP_006247418.1 | 2 - 3 | 749 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt28
Genomic Map