Assay Details
| Gene Symbol: Krt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008804.1 | NP_001008804.1 | 5 - 6 | 1150 | 158 | |
| XM_006247391.2 | XP_006247453.1 | 5 - 6 | 7047 | 158 | ||
| XM_008768091.1 | XP_008766313.1 | 5 - 6 | 7047 | 158 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Krt10
Genomic Map