Assay Details
| Gene Symbol: Krt85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008811.1 | NP_001008811.1 | 3 - 4 | 690 | 101 | |
| XM_006242351.2 | XP_006242413.1 | 3 - 4 | 746 | 101 | ||
| GenBank mRNA | AB355640.1 | - | 3 - 4 | 690 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt85
Genomic Map