Assay Details
| Gene Symbol: Krt85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008811.1 | NP_001008811.1 | 6 - 7 | 1075 | 105 | |
| XM_006242351.2 | XP_006242413.1 | 6 - 7 | 1131 | 105 | ||
| GenBank mRNA | AB355640.1 | - | 6 - 7 | 1075 | 105 | |
| Gene Symbol: Krt83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101675.1 | NP_001095145.1 | 6 - 7 | 1039 | 105 | |
| GenBank mRNA | AB354637.1 | - | 6 - 7 | 1039 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krt85
Genomic Map