Assay Details
| Gene Symbol: Fhl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013172.1 | NP_001013190.1 | 1 | 1450 | 115 | |
| XM_006241179.2 | XP_006241241.1 | 2 | 1808 | 115 | ||
| GenBank mRNA | BC078961.1 | - | 1 | 1450 | 7111014 | 115 |
| BC099213.1 | - | 2 | 1782 | 7384591 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fhl4
Genomic Map