Assay Details
| Gene Symbol: Prp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013211.1 | NP_001013229.1 | 2 - 3 | 858 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P090503 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P090508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173635 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P090508 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P090503 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P090503 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P090508 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P090503 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P090508 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P318572 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39089 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14644 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71672 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5425 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101333 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4145 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35703 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42315 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47487 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10859189 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859188 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10859190 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10859187 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400013758.rn.1 | TC0400001896.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400013759.rn.1 | TC0400001896.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400013761.rn.1 | TC0400001896.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400013760.rn.1 | TC0400001896.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400013757.rn.1 | TC0400001896.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400013756.rn.1 | TC0400001896.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400013764.rn.1 | TC0400001896.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10859187 | - | - |
| Rat Genome U34 Set | M64793_at | - | - |

Prp2
Genomic Map