Assay Details
| Gene Symbol: Phf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024272.1 | NP_001019443.1 | 4 - 5 | 413 | 74 | |
| GenBank mRNA | BC088433.1 | - | 4 - 5 | 413 | 7124805 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|

Phf11
Genomic Map