Assay Details
| Gene Symbol: Phf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024272.1 | NP_001019443.1 | 5 - 6 | 468 | 87 | |
| GenBank mRNA | BC088433.1 | - | 5 - 6 | 468 | 7124805 | 87 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Phf11
Genomic Map