Assay Details
| Gene Symbol: Apcdd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235711.2 | XP_006235773.1 | 3 - 4 | 1238 | 98 | |
| XM_008762580.1 | XP_008760802.1 | 3 - 4 | 3320 | 98 | ||
| GenBank mRNA | BC081904.1 | - | 1078 | 7110948 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P148396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6437 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P148396 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P148396 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P148396 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76177 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4234 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42023 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102547 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36919 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33145 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27389 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34447 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10852161 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852166 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852165 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852167 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10852164 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300037768.rn.1 | TC0300004960.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300037769.rn.1 | TC0300004960.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10852160 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365411 | - | - |

Apcdd1l
Genomic Map