Assay Details
| Gene Symbol: Tas2r138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024685.1 | NP_001019856.1 | 1 | 945 | 84 | |
| GenBank mRNA | AY916512.1 | - | 1 | 945 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tas2r138
Genomic Map