Assay Details
| Gene Symbol: Rsph6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024748.1 | NP_001019919.1 | 2 - 3 | 911 | 82 | |
| XM_008758909.1 | XP_008757131.1 | 2 - 3 | 966 | 82 | ||
| GenBank mRNA | BC097421.1 | - | 2 - 3 | 911 | 7454827 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Rsph6a
Genomic Map