Assay Details
| Gene Symbol: Osgepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024787.1 | NP_001019958.1 | 2 - 3 | 350 | 59 | |
| XM_006244893.2 | XP_006244955.1 | 2 - 3 | 399 | 59 | ||
| XM_006244894.2 | XP_006244956.1 | 2 - 3 | 363 | 59 | ||
| GenBank mRNA | BC078974.1 | - | 2 - 3 | 370 | 59 | |
| BC097950.1 | - | 2 - 3 | 350 | 7389045 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osgepl1
Genomic Map