Assay Details
| Gene Symbol: LOC301165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025662.2 | NP_001020833.2 | 4 - 5 | 2545 | 83 | |
| GenBank mRNA | BC097439.1 | - | 4 - 5 | 2545 | 7455751 | 83 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC301165
Genomic Map