Assay Details
| Gene Symbol: Cmtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029914.1 | NP_001025085.1 | 1 - 2 | 840 | 76 | |
| GenBank mRNA | BC098029.1 | - | 1 - 2 | 840 | 7456234 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm1
Genomic Map