Assay Details
| Gene Symbol: Paqr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034081.1 | NP_001029253.1 | 1 | 879 | 110 | |
| XM_006239130.2 | XP_006239192.1 | 2 | 977 | 110 | ||
| XM_008764145.1 | XP_008762367.1 | 3 | 1200 | 110 | ||
| XM_008764146.1 | XP_008762368.1 | 3 | 988 | 110 | ||
| GenBank mRNA | DQ027002.1 | - | 1 | 879 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Paqr7
Genomic Map