Assay Details
| Gene Symbol: Odf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024126.5 | NP_077040.2 | 1 - 2 | 353 | 88 | |
| GenBank mRNA | BC078708.1 | - | 1 - 2 | 353 | 7114007 | 88 |
| M58677.1 | - | 885 | 87 | |||
| M88759.1 | - | 1 - 2 | 370 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Odf1
Genomic Map