Assay Details
| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175604.2 | NP_783194.2 | 1 - 2 | 402 | 98 | |
| GenBank mRNA | BC088428.1 | - | 1 - 2 | 367 | 98 | |
| BC099797.1 | - | 1 - 2 | 402 | 7936110 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Yrdc
Genomic Map