Assay Details
| Gene Symbol: Cyp11b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012537.3 | NP_036669.3 | 6 - 7 | 1116 | 151 | |
| GenBank mRNA | D11354.1 | - | 6 - 7 | 1116 | 151 | |
| X15431.1 | - | 6 - 7 | 1116 | 151 | ||
More Information
Set Membership: |
|

Cyp11b1
Genomic Map