Assay Details
| Gene Symbol: Abcg3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037205.1 | NP_001032282.1 | 3 - 4 | 332 | 154 | |
| GenBank mRNA | BC100073.1 | - | 3 - 4 | 332 | 7372627 | 154 |
| Gene Symbol: LOC103690063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769965.1 | XP_008768187.1 | 3 - 4 | 904 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcg3l4
Genomic Map