Assay Details
| Gene Symbol: Aste1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766544.1 | XP_008764766.1 | 4 - 5 | 2390 | 109 | |
| GenBank mRNA | BC083913.1 | - | 5 - 6 | 753 | 109 | |
More Information
Set Membership: |
|

Aste1
Genomic Map