Assay Details
| Gene Symbol: Itpripl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025043.1 | NP_001020214.1 | 2 | 1770 | 87 | |
| XM_006234927.2 | XP_006234989.1 | 2 | 2283 | 87 | ||
| GenBank mRNA | BC082013.1 | - | 2 | 1770 | 7132065 | 87 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Itpripl1
Genomic Map