Assay Details
| Gene Symbol: Mrps28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047909.1 | NP_001041374.1 | 2 - 3 | 383 | 82 | |
| GenBank mRNA | AY383686.1 | - | 2 - 3 | 383 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps28
Genomic Map