Assay Details
| Gene Symbol: Mfsd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047910.1 | NP_001041375.1 | 5 - 6 | 671 | 68 | |
| GenBank mRNA | AY325200.1 | - | 5 - 6 | 671 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd8
Genomic Map