Assay Details
| Gene Symbol: Gpr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001071777.1 | NP_001065245.1 | 1 | 963 | 75 | |
| XM_006254604.2 | XP_006254666.1 | 2 | 2480 | 75 | ||
| GenBank mRNA | DQ777767.1 | - | 1 | 963 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr17
Genomic Map