Assay Details
Gene Symbol: RGD1562638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100944.1 | NP_001094414.1 | 3 - 4 | 2530 | 140 | |
XM_008771316.1 | XP_008769538.1 | 7 - 8 | 3336 | 140 | ||
XM_008771317.1 | XP_008769539.1 | 6 - 7 | 3284 | 140 | ||
XM_008771318.1 | XP_008769540.1 | 8 - 9 | 3390 | 140 | ||
XM_008771319.1 | XP_008769541.1 | 7 - 8 | 3336 | 140 | ||
XM_008771320.1 | XP_008769542.1 | 5 - 6 | 3550 | 140 | ||
XM_008771321.1 | XP_008769543.1 | 4 - 5 | 2963 | 140 | ||
XM_008771322.1 | XP_008769544.1 | 7 - 8 | 3336 | 140 | ||
GenBank mRNA | BC128761.1 | - | 3 - 4 | 2530 | 7113007 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
