Assay Details
Gene Symbol: MT-CO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | NC_001665.COX1.0 | - | 1241 | 94 |

More Information
Additional Information:
NC_001665 (strand:+; 5323..6867); mitochondrially encoded cytochrome c oxidase I
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
