Assay Details
| Gene Symbol: MT-ND4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | NC_001665.ND4L.0 | - | 149 | 130 | ||
More Information
Additional Information:
NC_001665 (strand:+; 9870..10166); mitochondrially encoded NADH 4L
Set Membership: |
|

MT-ND4L
Genomic Map