Assay Details
Gene Symbol: MT-CO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | NC_001665.COX3.0 | - | 446 | 142 |

More Information
Additional Information:
NC_001665 (strand:+; 8599..9382); mitochondrially encoded cytochrome c oxidase III
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
