Assay Details
| Gene Symbol: Efs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106033.2 | NP_001099503.1 | 3 - 4 | 794 | 90 | |
| GenBank mRNA | BC161942.1 | - | 3 - 4 | 794 | 9023909 | 90 |
More Information
Updated Mapping Information
Set Membership: |
|

Efs
Genomic Map