Assay Details
| Gene Symbol: Cttnbp2nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107712.2 | NP_001101182.2 | 5 - 6 | 578 | 136 | |
| XM_006233087.1 | XP_006233149.1 | 4 - 5 | 555 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P819787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69426 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P819787 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P819787 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P819787 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P459422 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94748 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10825705 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825706 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825707 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10825702 | - | - |
| Rat Genome 230 2.0 Array | 1379450_at | - | - |
| Rat Genome 230 2.0 Array | 1379450_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382873_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382873_at | - | - |
| Affymetrix HT Rat Focus Array | 1382873_at | - | - |
| Affymetrix HT Rat Focus Array | 1379450_at | - | - |
| Affymetrix HT Rat Focus Array | 1382873_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379450_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200030308.rn.1 | TC0200004389.rn.1 | RTA TC |
| Rat Expression Set 230 | 1379450_at | - | - |
| Rat Expression Set 230 | 1379450_PM_at | - | - |
| Rat Expression Set 230 | 1382873_PM_at | - | - |
| Rat Expression Set 230 | 1382873_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825702 | - | - |
| Rat Genome U34 Set | rc_AI234152_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21527592 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353445 | - | - |

Cttnbp2nl
Genomic Map