Assay Details
| Gene Symbol: Rin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108052.1 | NP_001101522.1 | 3 - 4 | 439 | 67 | |
| XM_006240473.2 | XP_006240535.1 | 3 - 4 | 708 | 67 | ||
| XM_006240474.2 | XP_006240536.1 | 4 - 5 | 496 | 67 | ||
| XM_008764804.1 | XP_008763026.1 | 3 - 4 | 5086 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rin3
Genomic Map