Assay Details
| Gene Symbol: H2afj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109610.2 | NP_001103080.1 | 1 | 447 | 78 | |
| GenBank mRNA | BC157816.1 | - | 1 | 447 | 9034669 | 78 |
| Gene Symbol: LOC103690002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763447.1 | XP_008761669.1 | 1 | 486 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

H2afj
Genomic Map