Assay Details
| Gene Symbol: Crygc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033483.1 | NP_264077.1 | 1 - 2 | 260 | 63 | |
| GenBank mRNA | BF523642.1 | - | 1 - 2 | 266 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crygc
Genomic Map