Assay Details
| Gene Symbol: Ang | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006992.1 | NP_001006993.1 | 1 | 143 | 150 | |
| XM_006251874.2 | XP_006251936.1 | 2 | 326 | 150 | ||
| GenBank mRNA | AY395869.1 | - | 1 | 143 | 150 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ang
Genomic Map