Assay Details
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leucine rich repeat containing 4C | Lrrc4c | RGD1311013 | -1.2mb | Sense | Chr. 3 - 86586354 - 86819922 on NCBI Build Rnor_6.0 |
Downstream | uncharacterized LOC102551620 | LOC102551620 | 28.8kb | Anti-sense | Chr. 3 - 88030140 - 88035306 on NCBI Build Rnor_6.0 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
