Assay Details
| Gene Symbol: LOC287004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110638.1 | 3 - 4 | 851 | 126 | ||
| NR_110640.1 | 3 - 4 | 851 | 126 | |||
| NR_110641.1 | 1 - 2 | 427 | 126 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein unc-80 homolog | LOC103693241 | -41.5kb | Sense | Chr. 9 - 72663790 - 72665685 on NCBI Build Rnor_6.0 | |
| Downstream | tensin 1 | Tns1 | Tns | 505.1kb | Sense | Chr. 9 - 81191792 - 81401027 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC287004
Genomic Map